Lazy-L-Spreaders™ Microbiological Spreaders, Excel Scientific
490007-358
: Avantor
- Description:Lazy-L-Spreaders, Individually Wrapped, Sterile
- Cat. no.:490007-358
- Supplier No.:SPR-L-S01
Lazy-L-Spreaders disposable microbiological spreaders provide an economical alternative to bending glass rods or pipets for spreading samples on agar surfaces and filters in Petri dishes. They are supplied sterile -- no need to flame sterilize or autoclave.
- Unlike glass spreaders, durable polystyrene Lazy-L-Spreaders are non-breakable and pose no safety hazard
- L-shaped for convenient "lazy" spreading; just turn the petri dish for smooth, even sample distribution
- Smooth spreading surface and a slight upward turn reduce the chance that the spreader will dig into agar
Packaged in pre-sterilized pouches of one or 10 each
Pre-sterilization eliminates flaming or autoclaving and reduces risk of contamination
No more time spent bending glass rods or waiting for glass or metal spreadersto cool before use.