Order Entry
Mexico
ContactUsLinkComponent
GeneMate UltraFlux® I, 8-Strip Standard PCR Tubes with Individually Attached, Optically-Clear Flat Caps, Scientific Specialties
GeneMate UltraFlux® I, 8-Strip Standard PCR Tubes with Individually Attached, Optically-Clear Flat Caps, Scientific Specialties
  490003-726
undefined
GeneMate UltraFlux® I, 8-Strip Standard PCR Tubes with Individually Attached, Optically-Clear Flat Caps, Scientific Specialties
  490003-726
 :  3248-00-210IS
  • Capacity:
    0.1 ml
  • Color:
    Natural
  • Description:
    8-Strip tubes, LP
  • Cat. no.:
    490003-726
  • Packaging:
    120 Strips of 8
  • Supplier No.:
    3248-00-210IS
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR) and standard PCR. Secure evaporation-resistant seal between cap lid and tube. Individual tubes have caps closing at an angle.
  • Angled caps prevent interference from adjacent tubes in thermal cycler
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Robust interwell linkages makes tubes more rigid and sturdy
  • Optically-clear flat caps
  • Crystal-clear clarity, easy to visualize content of each well
Available in full range of colors or white (enhances qPCR signals).
Certified free DNase, RNase, and detectable DNA.
 : GeneMate 96-well PCR tube storage racks are available separately.