Order Entry
Mexico
ContactUsLinkComponent
MBP 0.1 to 10 µl Graduated Ultra Micro Pipette Tips, Thermo Scientific
MBP 0.1 to 10 µl Graduated Ultra Micro Pipette Tips, Thermo Scientific
  490012-916
undefined
MBP 0.1 to 10 µl Graduated Ultra Micro Pipette Tips, Thermo Scientific
  490012-916
 :  104
  • Volume:
    0.1 - 10 µl
  • Packed:
    Bagged
  • Cat. no.:
    490012-916
  • Sterile:
    -
  • Tip Style:
    Ultra Micro
  • Supplier No.:
    104
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product
  • Natural
  • Metal-free tips: All materials used in the manufacture of these tips are free of trace metals
  • Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Pipetman®, Biohit Proline®, Eppendorf®, Jencons Sealpette®, Labpette™, Nichiryo, Oxford-Benchmate®, Rainin, Socorex®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non sterilized product is not certified as pyrogen-free.)
 : Certificate of analysis and Certificate of irradiation are available upon request.