Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Balance Curved Forceps
VWR® Balance Curved Forceps
Stainless steel, 102 mm (4")
Catalog # 89259-950
Supplier:   VWR International
VWR® Balance Curved Forceps
Catalog # 89259-950
Supplier:  VWR International
VWR® Balance Curved Forceps
VWR® Balance Curved Forceps
Stainless steel, 102 mm (4")
Price
$81.73
Log in for availability
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents
VWR® Balance Forceps are curved with plastic tips.
  • Stainless steel construction
Curved forceps are designed for precise handling of fine objects, built from non-corrosive stainless steel and featuring inert plastic tips.
Documents
Specifications
  • Length:
    102 mm (4")
  • Material:
    Stainless steel