Order Entry
Mexico
ContactUsLinkComponent
Fluorescence Microscopy Immersion Oil, Electron Microscopy Sciences
  100501-602
Minority or Woman-Owned Business Enterprise
 :  
undefined
Fluorescence Microscopy Immersion Oil, Electron Microscopy Sciences
  100501-602
 :  16916-01
Minority or Woman-Owned Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Immersion Oil Type FF
  • Size:
    1 oz.
  • Cat. no.:
    100501-602
  • Supplier No.:
    16916-01
Two new PCB-free immersion oils are availablefor fluorescence microscopy.
  • FF has virtually no background fluorescence
  • Extremely low fluorescence by LDF
  • Low fluorescence by HF
New Immersion oil HF provide low fluorescence and the correct optical properties and FF has irtually no fluorescence but slightly less than ideal optical properties. Applicator bottles have a glass ball rod in the cap.