Order Entry
Mexico
ContactUsLinkComponent
Liqui-nox® Phosphate Free Anionic and Non-Ionic Detergents, Electron Microscopy Sciences
Liqui-nox® Phosphate Free Anionic and Non-Ionic Detergents, Electron Microscopy Sciences
Catalog # 100503-404
Minority or Woman-Owned Business Enterprise
undefined
Liqui-nox® Phosphate Free Anionic and Non-Ionic Detergents, Electron Microscopy Sciences
Catalog # 100503-404
Supplier Number:  60515
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Liqui-nox® Phosphate Free Anionic and Non-ionic Detergent
  • Size:
    0.25 gal.
  • Cat. no.:
    100503-404
  • Packaging:
    Bottle
  • Supplier No.:
    60515
Specifications
About this item
Phosphate free anionic and non-ionic liquid detergents for manual and ultrasonic cleaning with mild pH.