Loctite Adhesive, Electron Microscopy Sciences
Product Details & Documents
A variety of Loctite adhesives are available that will bond a range of materials.
- 409™ Super Bonder®
- 454™ Prism®
- 861™ Prism®
- 349™ Impruv®
- 4011™ Prism®
The 409™ Super Bonder® is a general-purpose gel adhesive, clear, gap fills 0.010”; surface insensitive Ethyl, tensile shear strength 3,200 psi, and temperature range: –65°F to 180°F
The 454™ Prism® is for use with porous surfaces, clear, gap fills 0.010”; surface insensitive Ethyl, gel type, tensile shear strength 3200 psi, temperature range: –65°F to 180°F. Cure speed: fixture: 15 seconds, full: 24 hours.
The 861™ Prism® is for use with flexible surfaces, clear, gap fill 0.008”; surface flexible Alkyl, viscosity 4,000 cP, tensile shear strength 2465 psi, temperature range: –65°F to 212°F. Cure speed: fixture: 25 seconds, full: 24 hours.
The 349™ Impruv® is for bonding glass/metal; appearance: clear/straw, cure type: UV, viscosity: 9,500cP, shore hardness: D70, temperature range: –65°F to 266°F.
Cure speed: fixture: 75 seconds, full: 24 hours.
Specifications
Specifications
- Description:Loctite Prism 4011
- Size:20 g