Order Entry
Mexico
Orders LinkContactUsLinkComponent
Glycine for synthesis, Sigma-Aldrich®

Glycine for synthesis, Sigma-Aldrich®

Plastic bottle, 250 g
Catalog # EM8.16013.0250
Supplier #:  8.16013.0250
Glycine for synthesis, Sigma-Aldrich®
Catalog # EM8.16013.0250
CAS Number:  56-40-6
Glycine for synthesis, Sigma-Aldrich®
Glycine for synthesis, Sigma-Aldrich®

Plastic bottle, 250 g

CAS Number:  56-40-6
Price
$1,840.83
Log in for availability
Each (250g) Case de 15 (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  •  :M-Clarity™ quality level = MQ200

Documents

Specifications

Specification Test Results

  • Solubility:
    225 g/l (20 °C)
  • Melting Point:
    232 - 236 °C (decomposition)
  • Molar Mass:
    75.06 g/mol
  • Bulk Density:
    920 kg/m3
  • Density:
    1.595 g/cm3 (20 °C)
  • pH:
    5.9 - 6.4 (50 g/l, H2O, 20 °C)

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.