Photometer NOVA 60 A Spectroquant®
EM1.09752.0001
- Description:Spectroquant® NOVA 60A Photometer
- Environmentally Preferable:
- Cat. no.:EM1.09752.0001
- Supplier No.:1.09752.0001
The Spectroquant® NOVA 60 A are suitable for the routine analysis of all water types and capable of measuring all the ready-to-use cell tests as well as reagent tests.
- Barcode reading of all Spectroquant® tests
- Automatic selection of method and presentation of result
- Integrated QA/QC checks - Three modes, checking instrument, the method and sample volume
- Absorbance, concentration, transmission measuring modes
Designed for some critical tests, measurement to ppb levels, such as cyanide.