Order Entry
Mexico
ContactUsLinkComponent
n-Pentane EMSURE®
Flammable
Health
Irritant
Environment
n-Pentane EMSURE®
Catalog # EM1.07177.2500
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Pentane EMSURE®
Catalog # EM1.07177.2500
Supplier Number:  1.07177.2500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. no.:
    EM1.07177.2500
  • Supplier No.:
    1.07177.2500

Specifications

Specification Test Results

  • Solubility:
    0.04 g/l (25 °C)
  • Melting Point:
    -130 °C
  • Molar Mass:
    72.15 g/mol
  • Boiling Point:
    36 °C (1013 hPa)
  • Vapor Pressure:
    570 hPa (20 °C)
  • Flash Point:
    -48 °C (refers to pure substance)
  • Refractive Index:
    1.358
  • Explosion Limit:
    1.4 - 8 %(V)
  • Density:
    0.63 g/cm3 (20 °C)
  • Ignition Point:
    285 °C

Specification Test Results

About this item

 :
 :72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Boiling Pt:35 - 36 °C (1013 hPa)
Melting Pt:–130 °C
Melting Pt:-130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
Flash Pt:-40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498