


N,N-Dimethylformamide EMSURE® ACS
Catalog # EM1.03053.2500
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. no.:EM1.03053.2500
- Supplier No.:1.03053.2500
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-61 °C
- Molar Mass:73.09 g/mol
- Boiling Point:153 °C (1013 hPa)
- Vapor Pressure:3.77 hPa (20 °C)
- Flash Point:58 °C
- Refractive Index:1.43
- Explosion Limit:2.2 - 16 %(V)
- Density:0.94 g/cm3 (20 °C)
- pH:7 (200 g/l, H2O, 20 °C)
- Ignition Point:410 °C
Specification Test Results
About this item
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (lit.)
Boiling Pt:153 °C (1013 hPa)
Melting Pt:−61 °C (lit.)
Melting Pt:-61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284