Order Entry
Mexico
ContactUsLinkComponent
Molybdic acid
Health
Irritant
Molybdic acid
Catalog # EM1.00400.1000
CAS Number:  
Health
Irritant
Molybdic acid
Catalog # EM1.00400.1000
Supplier Number:  1.00400.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg
  • Cat. no.:
    EM1.00400.1000
  • Supplier No.:
    1.00400.1000

Specifications

Specification Test Results

  • Assay (precipitative titration):
    ≥ 85 %
  • Chloride (Cl):
    ≤ 0.01 %
  • Phosphate, Arsenate, Silicate (as PO₄):
    ≤ 0.005 %
  • Sulfate (SO₄):
    ≤ 0.02 %
  • Fe (Iron):
    ≤ 0.005 %
  • NH₄ (Ammonium):
    about 6

Specification Test Results

About this item

 :
Merck Index:13,06262
Boiling Pt:∼300 °C (1013 hPa)
Boiling Pt:~ 300 °C (1013 hPa)
Melting Pt:300 °C
Density:3.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :161.95 g/mol
EINECS:231-970-5
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:7782-91-4
Formula:H₂MoO₄
MDL Number:MFCD00036287