SP Bel-Art Poxygrid® Conical Tube Rack, 15 ml, Bel-Art Products, a part of SP
Catalog # 10600-004
Supplier: Bel-Art Products, a Part of SP
Specifications
- No. of Holes:15
- Description:Poxygrid® 15mL Conical Tube Rack
- Cat. no.:10600-004
- L×W×H:12.7 x 8.3 x 7.6 cm (5 x 3¹/₄ x 3")
- Supplier No.:187220000
Specifications
About this item
Epoxy-coated steel wire rack holds 15 conical bottom 15 mL centrifuge tubes.
- Heat-cured epoxy coating
- Withstands extreme temperature range
- Suited for holding conical tubes
- Autoclavable
The rack is generously spaced for easy handling
Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids, and weak alkalis. It will withstand a temperature range of -223 to 121°C (-369 to 250°F), and can be autoclaved.
Top and middle sections have 18 mm square holes and are centered over rails in the base that hold the tips of the tubes.