Simple Machines, Pulley Model
Catalog # 470210-810
Supplier: EISCO SCIENTIFIC LLC
Specifications
- Description:Simple Machines, Pulley Model
- Cat. no.:470210-810
- Supplier No.:WDMS12
Specifications
About this item
This wooden machine has a sturdy wooden construction ideal for classroom use.
- Demonstrate the use of pulleys
- Includes manual
This apparatus can model how the use of several pulleys increases one's mechanical advantage. Students can use this both qualitatively and quantitatively and its design lends itself easily to inquiry based activities. Includes an instruction manual to facilitate student and teacher use of the apparatus.