R2A Agar, Hardy Diagnostics
R2A Agar, 15x100mm, 18 ml, Plate
Supplier: HARDY DIAGNOSTICS
Supplier #: G54
R2A Agar, Hardy Diagnostics
Catalog # 89410-652
Supplier: HARDY DIAGNOSTICS
Product available on GSA Advantage®
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
R2A Agar, 15x100mm plate, order by the package of 10
For the detection of heterotrophic bacteria in water samples.
Documents
Certificates
Search by Lot NumberHardy Diagnostics Cosmetics and Personal Care Microbiology
Product Literature
Specifications
Specifications
- Pack type:Plate
- Description:R2A Agar
- Size:15x100mm
- Fill Volume:18 ml
- Application:For the detection of heterotrophic bacteria in water samples.
- Cat. No.:89410-652
- Storage temperature:2-8C



