Eisco® Skull Model
Skull, 3, 20 cm, 20 cm, 15 cm
Catalog # 470212-482
Supplier: EISCO SCIENTIFIC LLC
Supplier #: AMCH1004AS
Jump to:
Product Details & Documents
This life-size human skull shows all of the details of an adult human skull.
- Life Size
- Moveable Lower Jaw
- Removable Skull Cap
This realistic life-size human skull shows all of the bones and major structures of an adult human skull, including the fusions between bones and cartilaginous regions.
Documents
Specifications
- Description:Skull
- Height:15 cm
- Length:20 cm
- Width:20 cm
- No. of Parts:3
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



