Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Eisco® Dissectable Eye

Dissectible Eye, 6, 13 cm, 13 cm, 23 cm
Catalog # 470212-468
 :  AM34AS
Eisco® Dissectable Eye
Catalog # 470212-468
Eisco® Dissectable Eye
Eisco® Dissectable Eye

Dissectible Eye, 6, 13 cm, 13 cm, 23 cm

Price
$2,535.51
Log in for availability
Each (1UN)
Product Details & Documents

The six-part human eye model (magnified 5×) is a very highly detailed demonstration of the human eye, allowing students to deconstruct the eyeball into six smaller parts to examine the major internal structures.

Specifications
  • Description:
    Dissectible Eye
  • Height:
    23 cm
  • Length:
    13 cm
  • Width:
    13 cm
  • No. of Parts:
    6