Order Entry
Mexico
ContactUsLinkComponent
API® 20 Microbial Identification Kits, bioMerieux
API® 20 Microbial Identification Kits, bioMerieux
Catalog # 95060-670
CAS Number:  
API® 20 Microbial Identification Kits, bioMerieux
Catalog # 95060-670
Supplier Number:  20050
CAS Number:  
Specifications
  • Description:
    API® 20 NE Microbial Identification Kit
  • Cat. no.:
    95060-670
  • Supplier No.:
    20050
Specifications
About this item
Standardized, miniature version of conventional methods conducting various microbial tests.
  • Standardized
  • Easy to use
  • Identify gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other microorganisms
These microbial identification kits are used for identifying gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other clinically significant microorganisms.
Additional reagents required see package insert for details
https://us.vwr.com/store/product/18630970/microbial-identification-reagents-biomerieux
API Web software required for Identification -purchase directly from bioMerieux.