Order Entry
Mexico
ContactUsLinkComponent
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Catalog # 89499-726
undefined
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Catalog # 89499-726
Supplier Number:  T339-7S
Specifications
  • Sterility:
    Sterile
  • Volume:
    2.0 ml
  • Cat. no.:
    89499-726
  • Style:
    Conical Bottom
  • Graduations:
    Non graduated
  • Supplier No.:
    T339-7S
Specifications
About this item
MICREWTUBE® is ideal for freezer storage, boiling application and centrifugation.
The cap is molded with a deep internal lip that fits snugly against the interior wall of the tube thus preventing the contents from coming in contact with the seal or threads. In the silicone O-ring version, tubes and caps are made of polypropylene. Will withstand temperatures from –196 to +121 °C and are autoclavable. In the lip seal version, tubes are made of polypropylene and caps are made of high density polyethylene. Will withstand temperatures from –196 to +110 °C.
All tubes and closures are manufactured in a clean environment. Tubes and screw caps are made of heavy wall construction and are built to last. Non skirted tubes withstand high speed centrifugation up to 20000×g, skirted tubes up to 17000×g.
and will fit most standard microcentrifuge rotors. Six styles of caps to choose from and three sizes of conical bottom or self-standing tubes (0.5, 1.5 and 2 ml) are available in sterile and non sterile format. All sterile tubes are gamma irradiated and packaged in tamperproof, resealable bags to protect remaining tubes from contamination.
Certifications: Certified RNase, DNase, DNA and Pyrogen free.
Caution: Do not use Micrewtubes for storage in the liquid phase of liquid nitrogen. Such use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up resulting in possible explosion or biohazard release. Use appropriate safety procedures when handling and disposing of vials.