Order Entry
Mexico
Orders LinkContactUsLinkComponent
Phthalic acid for synthesis, Sigma-Aldrich®

Phthalic acid for synthesis, Sigma-Aldrich®

Plastic bottle, 1 kg
Catalog # EM8.22298.1000
Irritant
Supplier #:  8.22298.1000
Phthalic acid for synthesis, Sigma-Aldrich®
Catalog # EM8.22298.1000
CAS Number:  88-99-3
Phthalic acid for synthesis, Sigma-Aldrich®

Plastic bottle, 1 kg

CAS Number:  88-99-3
Price
$5,996.07
Log in for availability
Each (1kg) Case de 6 (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Pathalic acid
  • CAS Number:88-99-3
  • Formula:C₈H₆O₄
  • MDL Number:MFCD00002467
  • Molecular Weight:166.13 g/mol
  • Merck Index:12,07527
  • Boiling Pt:315 - 317 °C (1013 hPa)
  • Melting Pt:79 °C
  • Density:1.576 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Boiling Pt:315…317 °C (1013 hPa)
  • Storage Temperature:Ambient
  • EINECS:201-873-2

Documents

Specifications

Specification Test Results

  • Assay (acidimetric):
    ≥ 99.0 %
  • Identity (IR):
    passes test

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.