Order Entry
Mexico
ContactUsLinkComponent
Hydrazinium hydroxide (60 - 85%) in aqueous solution 80%, in water for synthesis
Toxic
Corrosive
Health
Environment
Hydrazinium hydroxide (60 - 85%) in aqueous solution 80%, in water for synthesis
Catalog # EM8.04604.1000
CAS Number:  
Toxic
Corrosive
Health
Environment
Hydrazinium hydroxide (60 - 85%) in aqueous solution 80%, in water for synthesis
Catalog # EM8.04604.1000
Supplier Number:  8.04604.1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. no.:
    EM8.04604.1000
  • Supplier No.:
    8.04604.1000

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -60 °C
  • Boiling Point:
    117 - 119 °C
  • Vapor Pressure:
    13 hPa (20 °C)
  • Flash Point:
    73 °C
  • Explosion Limit:
    4.7 - 99 %(V) (refers to pure substance)
  • Density:
    1.02 g/cm3 (20 °C)
  • pH:
    12 (510 g/l, H2O, 20 °C)
  • Ignition Point:
    310 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :32.05 g/mol (anhy)
Density:1.02 g/cm³ (20 °C)
UN:2030
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ200
EINECS:206-114-9
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:10217-52-4
 :7803-57-8
Formula:NH₂NH₂·xH₂O
MDL Number:MFCD00149931
ADR:8,II