
Sodium hydroxide concentrated aqueous solution 0.1 M Titrisol®
Catalog # EM1.09959.0001
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Pack type:Cardboard box
- Size:1 pack
- Cat. no.:EM1.09959.0001
- Supplier No.:1.09959.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.09 g/cm3 (20 °C)
- pH:13.8 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Boiling Pt:? 105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548
ADR:8,II