di-Sodium L(+)-tartrate dihydrate EMSURE®
Catalog # EM1.06663.1000
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. no.:EM1.06663.1000
- Supplier No.:1.06663.1000
Specifications
Specification Test Results
- Solubility:290 g/l (20 °C)
- Melting Point:57 °C
- Molar Mass:230.08 g/mol
- Bulk Density:460 kg/m3
- Density:1.82 g/cm3
- pH:8 (50 g/l, H2O, 25 °C)
Specification Test Results
About this item
:
:230.08 g/mol
Merck Index:12,08833
Melting Pt:57 °C
Density:1.818 g/cm³ (20 °C)
EINECS:212-773-3
CAS Number:6106-24-7
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035