Copper (II) chloride dihydrate EMSURE®
Catalog # EM1.02733.0250
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:250 g
- Cat. no.:EM1.02733.0250
- Supplier No.:1.02733.0250
Specifications
Specification Test Results
- Solubility:1150 g/l (20 °C)
- Melting Point:100 °C
- Molar Mass:170.48 g/mol
- Bulk Density:950 kg/m3
- Density:2.54 g/cm3 (20 °C)
- pH:3.0 - 3.8 (50 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS07
:GHS09
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674