Order Entry
Mexico
ContactUsLinkComponent
VACUBOY Hand Operator, INTEGRA Biosciences
VACUBOY Hand Operator, INTEGRA Biosciences
Catalog # 37001-682
Supplier:  INTEGRA BIOSCIENCE
undefined
VACUBOY Hand Operator, INTEGRA Biosciences
Catalog # 37001-682
Supplier:  INTEGRA BIOSCIENCE
Supplier Number:  155500
Specifications
  • Description:
    VACUBOY® Hand Operator Set, with Additional Parts
  • Cat. no.:
    37001-682
  • Supplier No.:
    155500
Specifications
About this item
The ergonomic VACUBOY® hand operator and its adapters provide the flexibility to aspirate liquids from a wide range of laboratory containers, including cell culture vessels and multi-well plates
The VACUBOY® is the perfect match to the VACUSAFE Aspiration System.
Ordering information: Supplied complete with hand piece (for holding adapters or pipettes), rubber adapter for Pasteur pipettes, single channel stainless steel adapter (40mm), and single channel adapter for disposable tips with ejector.