SCIENCEWARE® Heavy-Duty Biohazard Disposal Bags, Bel-Art
58949-646
- Capacity:40 - 50 gal.
- Color:Red
- Thickness:2 mil.
- Cat. no.:58949-646
- W×L:940×1220 mm (37×48")
- Supplier No.:131653748
Protect against dangerous tears and leaks with extra strong, high impact polypropylene biohazard disposal bags. Labeling displays standard biohazard warning symbol and precautionary protocols in English, French, German, and Spanish.
- Heavy-duty polypropylene construction
- Prominent biohazard warning symbol
- Impact- and tear-resistant design
Labeling displays standard biohazard warning symbol and precautionary protocols in English, French, German, and Spanish.
Extra heavy-duty bags are a 0.05 mm (2 mil) thick blended polymer, autoclavable to 135 °C (275 °F) and feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
: Meet or exceed the ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests.
: Extra large sizes available, call for quote.