Order Entry
Mexico
Orders LinkContactUsLinkComponent
Filtering Flask, Microscale, Glassco
Filtering Flask, Microscale, Glassco
Filtering Flask, Microscale, 25 ml
Catalog # 89426-772
Supplier #:  259.471.01
Filtering Flask, Microscale, Glassco
Catalog # 89426-772
Filtering Flask, Microscale, Glassco
Filtering Flask, Microscale, Glassco
Filtering Flask, Microscale, 25 ml
Price
$194.76
Log in for availability
Each (1UN)
Product Details & Documents
Flask top is tooled for a #2 neoprene filter adapter or #3 rubber stopper
Manufactured from Borosilicate 3.3 glass.
Documents
Specifications
  • Capacity:
    25 ml
  • Description:
    Filtering Flask, Microscale