Iron Test, MQuant®, MilliporeSigma
2,2'-Bipyridine, 3-10-25-50-100-250-500 mg/L Fe(II), 100 strips
Supplier: MERCK, S.A. DE C.V. SV
Supplier #: 1100040001
Iron Test, MQuant®, MilliporeSigma
Catalog # EM1.10004.0001
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Jump to:
Product Details & Documents
Method: Colorimetric with Test Strips 3 - 10 - 25 - 50 - 100 - 250 - 500 mg/l Fe2+ MQuant™ Iron Test.
The test strips offer a very simple means of determining iron in all aqueous media and on metal surfaces. They can thus be used to distinguish between ferrous and non-ferrous metals.
MilliporeSigma # 1.10004.0001 replaces and is identical to MilliporeSigma # 10004-1
Documents
Specifications
Specifications
- Chemical method:2,2'-Bipyridine
- Range:3-10-25-50-100-250-500 mg/L Fe(II)
- No. of Tests:100 strips