Order Entry
Mexico
ContactUsLinkComponent
Reflectoquant® Hydroxymethylfurfural (HMF) Test, MilliporeSigma
Reflectoquant® Hydroxymethylfurfural (HMF) Test, MilliporeSigma
Catalog # EM1.17952.0001
CAS Number:  
Reflectoquant® Hydroxymethylfurfural (HMF) Test, MilliporeSigma
Catalog # EM1.17952.0001
Supplier Number:  1179520001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Reflectoquant® Hydroxymethylfurfural (HMF) Test Method: Reflectometric with Test Strips
  • Environmentally Preferable:
  • Cat. no.:
    EM1.17952.0001
  • Range:
    1.0 - 60.0 mg/L
  • Supplier No.:
    1.17952.0001
  • No. of Strips:
    50
Specifications
About this item
Method: Reflectometric With Test Strips 1.0 - 60.0 mg/l Reflectoquant® HMF Test.
Reflectoquant® Hydroxymethylfurfural (HMF) is the first easy-to-use rapid test of HMF, an important quality parameter in thermally treated fruit juices and honey samples. Enzymatic reaction.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.