Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

3Dimensional, Fixed Tilt Platform Rotator, PS-3D, Grant Instruments

Fixed Tilt 3D Platform Rotator
  97025-564
 :  PS-3D
3Dimensional, Fixed Tilt Platform Rotator, PS-3D, Grant Instruments
  97025-564
3Dimensional, Fixed Tilt Platform Rotator, PS-3D, Grant Instruments
3Dimensional, Fixed Tilt Platform Rotator, PS-3D, Grant Instruments

Fixed Tilt 3D Platform Rotator

 
$26,780.95
   
Each (1UN)
 

The compact PS-3D variable speed, fixed angle 3D rotator is ideally suited for personal use

Gentle movement keeps fragile tissue intact during incubations. Can be used at 4 to 40°C (80% relative humidity, non-condensing), in an incubator or cold room.

It provides a smooth orbital motion for mixing commonly used vessls- culture flasks, dishes, boxes and tubes. The reliable motor produces regulated and reproducible rotating throughout the speed range.

 : CE marked.

 : Supplied with non-slip mat; optional dimpled mat is available separately.
Manufacturers 2 year warranty.

 
  • Description:
    Fixed Tilt 3D Platform Rotator
  • Cat. No.:
    97025-564
  • Platform dimensions:
    210 x 210 mm
  • Operating time:
    24 hrs. (continuous operation)
  • Tilt angle:
  • Speed:
    5 to 60 rpm
  • Electrical:
    12V
  • Max. load:
    1 kg
  • Operating temperature:
    4...40 °C