Order Entry
Mexico
ContactUsLinkComponent
PIG® Mercury Spill Vacuum Kit, New Pig
  56620-340
undefined
PIG® Mercury Spill Vacuum Kit, New Pig
  56620-340
 :  KIT330
  • Capacity:
    Up to 177.4 ml (6 oz.)
  • Cat. no.:
    56620-340
  • Supplier No.:
    KIT330
Vacuum pump allows you to quickly and effectively capture mercury spills for proper recycling or disposal
  • Scoop and spatula make it easier to gather scattered mercury droplets without unnecessary scattering or loss
  • Durable polyethylene carrying case keeps all necessary equipment in one place
  • No hazardous components to generate additional hazardous waste
No wheels. Not UN rated.
Includes collection vials and cleanup gear for up to 6 spills