Order Entry
Mexico
ContactUsLinkComponent
Spill-X-A® Acid-Neutralizing Adsorbent, New Pig
Spill-X-A® Acid-Neutralizing Adsorbent, New Pig
Catalog # 89205-854
undefined
Spill-X-A® Acid-Neutralizing Adsorbent, New Pig
Catalog # 89205-854
Supplier Number:  GEN322
Specifications
  • Description:
    Pail, 50 lbs.
  • Cat. no.:
    89205-854
  • Supplier No.:
    GEN322
Specifications
About this item
Neutralizes and solidifies most mineral and organic acids for safe and easy cleanup, including sulfuric, hydrochloric, nitric, and hydrofluoric
Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Excellent for use in shipping/receiving areas, acid container storage areas and acid use and transfer sites. Non-toxic, free-flowing dry chemical is environmentally friendly.