Model 445715 Big Digit Remote Probe Hygro-Thermometer, Extech
Supplier: Avantor
Certificates
About this item
The model 445715 Hygro-Thermometer with Humidity Alert and Remote Probe displays 2.5cm (1") digits on an extra-large LCD
The adjustable probe clips onto meter or extends to 45.7cm (18"). Unit features min./max. control, with reset function.
Certifications: CE marked.
Ordering information: Supplied complete with built-in stand, wall mount bracket, sensor, and 1 AAA battery. Unit also includes manufacturer's one-year warranty.
Frequently Bought Together