Order Entry
Mexico
Orders LinkContactUsLinkComponent
Hardness 3 Solution, 100 mL, Hach

Hardness 3 Solution, 100 mL, Hach

Hardness 3 Solution, 100 mL Marked Dropping Bottle, EDTA
Catalog # 89230-874
Supplier #:  42632
Hardness 3 Solution, 100 mL, Hach
Catalog # 89230-874
CAS Number:  
Hardness 3 Solution, 100 mL, Hach
Hardness 3 Solution, 100 mL, Hach

Hardness 3 Solution, 100 mL Marked Dropping Bottle, EDTA

CAS Number:  
Price
$772.05
Log in for availability
Each (1UN)
Product Details & Documents

EDTA titrant solution for hardness determination by drop-count titration. 0.03 N (0.015 M).

0.03 N (0.015 M).

Specifications

Specifications

  • Chemical method:
    EDTA
  • Description:
    Hardness 3 Solution
  • Size:
    100 mL Marked Dropping Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.