Total Hardness Test Kit, Model 5-B, Hach
Catalog # 97026-826
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:EDTA
- Description:Total Hardness Test Kit, Model 5-B
- Method:Drop count titration/ EDTA-bulk powder
- Case style:L
- Cat. no.:97026-826
- Range:1 - 30 gpg CaCO₃
- Supplier No.:145300
- No. of Tests:100
Specifications
About this item
Trust the original Hach test kit! Simple drop-count titration measures hardness as CaCO3 (1 gpg = 17.1 mg/L). Utilizes drop count titration with a range of 1-30 gpg.
Utilizes drop count titration - a simple, economical titration method
Utilizes drop count titration with a range of 1-30 gpg.