Order Entry
Mexico
ContactUsLinkComponent
DPD Free Chlorine Reagent Powder Pillows, Hach
DPD Free Chlorine Reagent Powder Pillows, Hach
Catalog # 97026-718
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
DPD Free Chlorine Reagent Powder Pillows, Hach
Catalog # 97026-718
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    DPD Free Chlorine Reagent Powder Pillows
  • Size:
    5 ml sample
  • Method:
    DPD
  • Cat. no.:
    97026-718
  • Range:
    up to 3.5 mg/L Cl₂
  • Supplier No.:
    1407728
  • No. of Tests:
    1000

Specifications

About this item

For determination of free chlorine by the DPD method.
For use with Hach's Chlorine Visual Test Kits.
Range: up to 3.5 mg/L Cl2.

  • Boric acid-free