Suspension Clamp
Suspension Clamp
470137-844
: CH0667
Takes the Heat
Use this suspension clamp to safely hold a thermometer or specimens over a hotplate. The clamp keeps objects a distance of 110mm from the rod and the hook can be adjusted to any angle.
- Description:Suspension Clamp