Order Entry
Mexico
Orders LinkContactUsLinkComponent
Boss Head, Right Angle
Boss Head, Right Angle
Boss Head - Right Angle, Powder Coated
Catalog # 470137-826
Supplier #:  CH0663A
Boss Head, Right Angle
Catalog # 470137-826
Boss Head, Right Angle
Boss Head, Right Angle
Boss Head - Right Angle, Powder Coated
Price
$191.36
Log in for availability
Each (1UN)
Product Details & Documents
Securely holds rods at a 90 degree angle.
  • Made of a die cast alloy, powder-coated finish for durability
  • Fixed design, holds rods at right angles to each other in a sturdy fashion
  • Features metal thumbscrews for adjustments
  • Measures approximately 11×3×3 cm
It accepts rods up to 16 mm in diameter at a right angle to one another.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Specifications
  • Description:
    Boss Head - Right Angle, Powder Coated