Order Entry
Mexico
ContactUsLinkComponent
Auto-Desiccator Cabinets, Ace Glass
  89054-070
Product available on GSA Advantage®
Small Business Enterprise
undefined
Auto-Desiccator Cabinets, Ace Glass
  89054-070
 :  6257-50
Product available on GSA Advantage®
Small Business Enterprise
  • Volume:
    1.9 cu. ft. (internal)
  • Description:
    Vertical Profile Electric Auto-Desiccator Cabinet, Clear
  • Electrical:
    120V
  • Exterior dimensions:
    34.1W×41.4D×51.8H cm (13.4×16.3×20.4"). Door:22.5W×41.7H cm (8.9×16.4")
  • Cat. no.:
    89054-070
  • Supplier No.:
    6257-50
This stackable, plastic Secador™ desiccator cabinet is perfect for cramped locations
Can be safely stacked up to two high. Cabinet offered clear or blue with clear door. With patented dehumidifying module—automatic desiccant regeneration and built-in hygrometer. Unit has three shelves, four shelf positions. Patented sealed construction—for air-tight, dust- and moisture-free storage. Two latches; provision for lock or tamper evident seal.