Hydrogen chloride (2.7 - 6 mol/l) in 2-propanol 5 - 6 M
:7647-01-0
:
:HCl
:MFCD00011324
:36.46 g/mol
:0.909
:11 °C (52 °F)
:231-595-7
:2920
:8,II
:GHS02
:GHS05
:GHS06
:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.