Order Entry
Mexico
Orders LinkContactUsLinkComponent
hydraflock-feature.jpg
HydraFlock® and PurFlock® Ultra™ Specimen Collection Devices
HydraFlock provides superior collection and a quick release of specimens, enhanced by its unique microstructure with multi-length fibers. PurFlock Ultra’s high purity features make it an ideal swab for rapid diagnostic tests.
HydraFlock® and PurFlock® Ultra™ Specimen Collection Devices | Avantor
A new era in flock swab technology! Specifically engineered for a wide range of biological specimens.
HydraFlock®’s three-dimensional microstructure design facilitates greater specimen collection. The multi-length fibers have a greater absorption area than traditional perpendicular nylon flock fibers for more rapid elution and maximal sample preservation. Ideal for a variety of clinical settings and diagnostic tests.
Engineered for purity and maximal cell yield with a small sample size, PurFlock Ultra® flocked swabs are ideal for PCR, molecular assays, DFA testing, forensic applications and direct antigen testing. Constructed from technology the results in higher purity than traditional nylon flocked swabs, they provide the greatest balance between purity and sample collection efficiency on the market. Ideal for various rapid diagnostic tests.
VWR-Flock-3316-002.jpg
A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.
VWR-Flock-3317.jpg
PurFlock Ultra™’s high purity characteristics makes it an ideal swab for various rapid diagnostic tests. An ideal choice in flock swabs with excellent absorption and elution of specimens when HydraFlock’s high levels of absorbency aren’t necessary.