Order Entry
Mexico
Orders LinkContactUsLinkComponent
Laser-Microdissection-900x59012.jpg
Laser Microdissection
Laser Microdissection (LMD, also known as Laser Capture Microdissection or LCM) enables users to isolate specific single cells or entire areas of tissue. Powered by a unique laser design and dynamic software, Leica LMD systems allow users to easily isolate Regions of Interest (ROI) from entire areas of tissue down to single cells or even subcellular structures such as chromosomes.
Leica - Laser Microdissection | Avantor
Product selection
Laser-Microdissection-400x495.jpg
Laser Microdissection (LMD) enables users to isolate specific single cells or entire areas of tissue for downstream genomic and proteomic analyses.