Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's Freeze Dried Specimens | Avantor
Ward's Freeze Dried Specimens
Cost-Effective, Chemical-Free Dissection
Completely chemical-free, these specimens offer the maximum value and flexibility for your dissection lessons. Prepared through a process of sublimation, all water is removed from the specimen, leaving it perfectly preserved for extended storage and easy to re-hydrate when you’re ready to use in your classroom.
  • Unlimited shelf-life in the dried state for long-lasting storage
  • Lighter package weight for cost-effective shipping
  • Easy to use – every package includes step-by-step instructions for re-hydration
  • Rehydrated specimens last for months in your classroom, ideal for extended lab use
  • Organs and tissues retain natural coloration
free-dried-preserved-200.jpg
Shop for all Ward's freeze dried specimens.