HENKE-JECT® Single Use Syringes, 2-Piece
Product Details & Documents
Disposable, two-piece, without needle, PP barrel and PE piston, latex- and silicone oil-free. Syringes are also available with especially designed oral tip, which prevents accidental Luer or hypodermic connection, in volumes of 5 or 10 ml. Incompatible with injection needles. Please enquire for details.
- Defined position of the plunger at volume '0' to feel when plunger is completely inserted
- No dead space for 1 ml insulin and tuberculin syringe
- Extended graduation for wider range of applications
- Positive safety stop to prevent accidental spills
Syringes are also available with especially designed oral tip, which prevents accidental Luer or hypodermic connection, in volumes of 5 or 10 ml. Incompatible with injection needles. Please enquire for details.
Packaging: Sterile versions are packed in individual blister strips. Non sterile versions are bulk packed.
Specifications
- 용량:2 ml
- 설명:Single-use syringe, Luer tip
- Seq:4