VWR Clean Room Sleeve Protector, Alliance, Gamma Sterilized, 24 Inch Long, Thumb Loop Style, Ultrasonic Welded Construction, 1 Pair Per Bag, 200 Bags Per Case, Made in USA 1 * 200 items
제품 번호 VWRI76491-712
공급사: VWR Collection
New Product
Frequently Bought Together