ContactUsLinkComponent
VWR Filter Combination, 30mm, PTFE 0.22um filter, # 17 hose, small port conversion cover, Sterile,1 set/bag,1 bag/box 1 * 1 items
제품 번호 VWRA734-3673
공급사:  VWR Collection
New Product
VWR Filter Combination, 30mm, PTFE 0.22um filter, # 17 hose, small port conversion cover, Sterile,1 set/bag,1 bag/box 1 * 1 items
제품 번호 VWRA734-3673
공급사:  VWR Collection
New Product
Frequently Bought Together