VWR®, Cryo coder cap inserts
Cap insert, Green
공급사: VWR Collection
Jump to:
Product Details & Documents
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple colour options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
사양
- Colour:Green
- 설명:Cap insert
- Seq:03