Fitting, Black Nylon, Elbow, Hose Barb Union, 3/32" ID; 10/Pk
Catalog # MFLX41518-50
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- 바브 크기/튜브 내부 직경 (포트 1):3/32"
- 바브 크기/튜브 내부 직경 (포트 2):3/32"
- 연결 유형:Barbed Union
- Flow pattern:Straight
- 설명:Fitting, Black Nylon, Elbow, Hose Barb Union, 3/32" ID; 10/Pk
- Seq:19
- Environmentally Preferable:
- Model:41518-50
- Sterilisation:Ethylene oxide (ETO) only
- Pk:10
- Fitting material:Black Nylon
Frequently Bought Together