Masterflex® Fitting, PVDF, Straight, Hose Barb Union, 1/16" ID; 10/PK
Catalog # MFLX40703-00
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
- Connection Type: 1/16" ID
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- 바브 크기/튜브 내부 직경 (포트 1):0,0625"
- 바브 크기/튜브 내부 직경 (포트 2):0,0625"
- 연결 유형:Barb
- Flow pattern:Straight
- 설명:Fitting, PVDF, Straight, Hose Barb Union, ¹⁄₁₆" ID; 10/PK
- Seq:20
- Environmentally Preferable:
- Model:40703-00
- Orifice dimensions:0,03125"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Pk:10
- Fitting material:PVDF
Frequently Bought Together