Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Unions, 3/8" ID
제품 번호 MFLX31209-57
이 제품에 관하여
Ensure a tight seal every time.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
- Connection Type: 3/8" ID; 1/EA
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
- 바브 크기/튜브 내부 직경 (포트 1):3/8"
- 바브 크기/튜브 내부 직경 (포트 2):3/8"
- 연결 유형:Barbed Union
- Flow pattern:Y
- 설명:Fitting, Stainless Steel, Y-Connector, Hose Barb Unions, 3/8" ID
- Seq:07
- Environmentally Preferable:
- Model:31209-57
- Orifice dimensions:0,28125"
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:1 year
- Pk:1
- Barb size/tubing int.Ø (port 3):3/8"
- Fitting material:Stainless Steel 316
Frequently Bought Together